missing translation for 'onlineSavingsMsg'
Learn More

N-Formyl Oxcarbazepine, TRC

Product Code. 30389429
Change view
Click to view available options
Quantity:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30389429 10 mg
30392562 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30389429 Supplier Toronto Research Chemicals Supplier No. TRCF70088010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1346601-76-0
Molecular Formula C16H12N2O3
Molecular Weight (g/mol) 280.28
Synonym N-Formyl-10,11-dihydro-10-oxo-5H-dibenz[b,f]azepine-5-carboxamide
SMILES O=C(NC=O)N(C1=C(C2)C=CC=C1)C3=C(C2=O)C=CC=C3

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C16H12N2O3/c19-10-17-16(21)18-13-7-3-1-5-11(13)9-15(20)12-6-2-4-8-14(12)18/h1-8,10H,9H2,(H,17,19,21)
Formula Weight 280.0848
Percent Purity >95%
Chemical Name or Material N-Formyl Oxcarbazepine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.