missing translation for 'onlineSavingsMsg'
Learn More

N-Formyl N-(des(boric acid)-des(methylbutyl)) Bortezomib, TRC

Product Code. 30401373
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30401373 25 mg 25mg
30394132 5 mg 5mg
30397026 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30401373 Supplier Toronto Research Chemicals Supplier No. TRCF70030525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Formyl N-(des(boric acid)-des(methylbutyl)) Bortezomib
Analyte or Component Names N-Formyl N-(des(boric acid)-des(methylbutyl)) Bortezomib
Shipping Condition Room Temperature
Molecular Formula C15H14N4O3
Synonym (S)-N-(1-Formamido-1-oxo-3-phenylpropan-2-yl)pyrazine-2-carboxamide
InChI Formula InChI=1S/C15H14N4O3/c20-10-18-14(21)12(8-11-4-2-1-3-5-11)19-15(22)13-9-16-6-7-17-13/h1-7,9-10,12H,8H2,(H,19,22)(H,18,20,21)/t12-/m0/s1
Solution Type Neat
SMILES O=CNC([C@H](CC1=CC=CC=C1)NC(C2=CN=CC=N2)=O)=O
IUPAC Name (S)-N-(1-formamido-1-oxo-3-phenylpropan-2-yl)pyrazine-2-carboxamide
Industry Type Pharmaceutical
Molecular Weight (g/mol) 298.3
Quantity 25 mg
Formula Weight 298.11
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.