missing translation for 'onlineSavingsMsg'
Learn More

N-Formyl-L-methionine, 95%, Thermo Scientific Chemicals

Catalog Number 15410219
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
4289-98-9
C6H11NO3S
177.22
MFCD00021033
PYUSHNKNPOHWEZ-YFKPBYRVSA-N
n-formyl-l-methionine, n-formylmethionine, for-met-oh, s-2-formamido-4-methylthio butanoic acid, formyl-l-methionine, formylmethionine, l-methionine, n-formyl, unii-ps9357b4xh, n-formyl methionine
439750
CHEBI:16552
(2S)-2-formamido-4-methylsulfanylbutanoic acid
CSCC[C@H](NC=O)C(O)=O
This item is not returnable. View return policy
4289-98-9
C6H11NO3S
177.22
MFCD00021033
PYUSHNKNPOHWEZ-YFKPBYRVSA-N
n-formyl-l-methionine, n-formylmethionine, for-met-oh, s-2-formamido-4-methylthio butanoic acid, formyl-l-methionine, formylmethionine, l-methionine, n-formyl, unii-ps9357b4xh, n-formyl methionine
439750
CHEBI:16552
(2S)-2-formamido-4-methylsulfanylbutanoic acid
CSCC[C@H](NC=O)C(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Formula Weight 177.22
Percent Purity 95%
Quantity 5 g
Physical Form Solid
Chemical Name or Material N-Formyl-L-methionine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'