missing translation for 'onlineSavingsMsg'
Learn More

N-Formyl-L-leucine, TRC

Product Code. 30394409
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30394409 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30394409 Supplier Toronto Research Chemicals Supplier No. TRCF7005601G
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 6113-61-7
Molecular Formula C7 H13 N O3
Synonym L-Leucine, N-formyl-, Leucine, N-formyl-, L- (8CI), N-Formyl-L-leucine, (2S)-2-Formamido-4-methylpentanoic acid, (S)-(+)-2-Formamido-4-methylpentanoic acid, N-Formyl-S-leucine, N-Formylleucine, NSC 334321
InChI Formula InChI=1S/C7H13NO3/c1-5(2)3-6(7(10)11)8-4-9/h4-6H,3H2,1-2H3,(H,8,9)(H,10,11)/t6-/m0/s1
SMILES CC(C)C[C@H](NC=O)C(=O)O
IUPAC Name (2S)-2-formamido-4-methylpentanoic acid
Molecular Weight (g/mol) 159.18
Quantity 1 g
Formula Weight 159.0895
Shipping Condition Room Temperature
Chemical Name or Material N-Formyl-L-leucine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.