missing translation for 'onlineSavingsMsg'
Learn More

N-Formyl-L-leucine (3S,4S,6S)-3-Hexyltetrahydro-2-oxo-6-undecyl-2H-pyran-4-yl Ester, Mikromol™

Product Code. 17919704
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Product Code. Quantity unitSize
17919704 25 mg 25mg
1 options
This item is not returnable. View return policy

Product Code. 17919704

Brand: Mikromol MM1219.080025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

N-Formyl-L-leucine (3S,4S,6S)-3-Hexyltetrahydro-2-oxo-6-undecyl-2H-pyran-4-yl Ester
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Formyl-L-leucine (3S,4S,6S)-3-Hexyltetrahydro-2-oxo-6-undecyl-2H-pyran-4-yl Ester
CAS 130676-65-2
Analyte or Component Names N-Formyl-L-leucine (3S,4S,6S)-3-Hexyltetrahydro-2-oxo-6-undecyl-2H-pyran-4-yl Ester
Shipping Condition Ice pack (-20°C)
Molecular Formula C29H53NO5
Certifications/Compliance ISO 9001
Synonym [3S-(3alpha,4alpha,6beta)]-N-Formyl-L-leucine 3-Hexyltetrahydro-2-oxo-6-undecyl-2H-pyran-4-yl Ester, Ro 40-4441, Orlistat impurity
SMILES CCCCCCCCCCC[C@H]1C[C@H](OC(=O)[C@H](CC(C)C)NC=O)[C@H](CCCCCC)C(=O)O1
Molecular Weight (g/mol) 495.73
Quantity 25 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.