missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Formyl-L-leucine (1S)-1-[(2R,3S)-3-Carboxy-2-hydroxynonyl]dodecyl Ester, Mikromol™
Description
N-Formyl-L-leucine (1S)-1-[(2R,3S)-3-Carboxy-2-hydroxynonyl]dodecyl Ester
Specifications
Specifications
| Chemical Name or Material | (2S,3R,5S)-5-[(N-Formyl-L-leucyl)oxy]-2-hexyl-3-hydroxyhexadecanoic acid |
| CAS | 130793-28-1 |
| Analyte or Component Names | (2S,3R,5S)-5-[(N-Formyl-L-leucyl)oxy]-2-hexyl-3-hydroxyhexadecanoic acid |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C29 H55 N O6 |
| Certifications/Compliance | ISO 9001 |
| Synonym | N-Formyl-L-leucine (1S)-1-[(2R,3S)-3-Carboxy-2-hydroxynonyl]dodecyl Ester, (2S,3R,5S)-5-[(N-Formyl-L-leucyl)oxy]-2-hexyl-3-hydroxyhexadecanoic Acid |
| InChI Formula | InChI=1S/C29H55NO6/c1-5-7-9-11-12-13-14-15-16-18-24(36-29(35)26(30-22-31)20-23(3)4)21-27(32)25(28(33)34)19-17-10-8-6-2/h22-27,32H,5-21H2,1-4H3,(H,30,31)(H,33,34)/t24-,25-,26-,27+/m0/s1 |
| SMILES | CCCCCCCCCCC[C@@H](C[C@@H](O)[C@H](CCCCCC)C(=O)O)OC(=O)[C@H](CC(C)C)NC=O |
| IUPAC Name | (2S,3R,5S)-5-[(2S)-2-formamido-4-methylpentanoyl]oxy-2-hexyl-3-hydroxyhexadecanoic acid |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?