missing translation for 'onlineSavingsMsg'
Learn More

N-Formyl Desloratadine, TRC

Product Code. 17984565
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17984565 1 g 1g
17942535 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17984565 Supplier Toronto Research Chemicals Supplier No. TRCF7003001G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 117810-61-4
Molecular Formula C20 H19 Cl N2 O
Molecular Weight (g/mol) 338.83
Synonym N-Formyldesloratadine, 8-Chloro-6,11-dihydro-11-(N-formyl-4-piperidinylidene)-5H-benzo[5,6]cyclohepta[1,2-b]pyridine
IUPAC Name 4-(8-chloro-5,6-dihydrobenzo[1,2]cyclohepta[2,4-b]pyridin-11-ylidene)piperidine-1-carbaldehyde
SMILES Clc1ccc2C(=C3CCN(CC3)C=O)c4ncccc4CCc2c1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C20H19ClN2O/c21-17-5-6-18-16(12-17)4-3-15-2-1-9-22-20(15)19(18)14-7-10-23(13-24)11-8-14/h1-2,5-6,9,12-13H,3-4,7-8,10-11H2
Formula Weight 338.1186
Percent Purity >95%
Chemical Name or Material N-Formyl Desloratadine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.