missing translation for 'onlineSavingsMsg'
Learn More

N-Formyl Amlodipine, TRC

Product Code. 30393593
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30393593 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30393593 Supplier Toronto Research Chemicals Supplier No. TRCF68040525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Formyl Amlodipine
CAS 93848-81-8
Molecular Formula C21 H25 Cl N2 O6
Quantity 25 mg
Synonym 3,5-Pyridinedicarboxylic acid, 4-(2-chlorophenyl)-2-[[2-(formylamino)ethoxy]methyl]-1,4-dihydro-6-methyl-, 3-ethyl 5-methyl ester, O3-Ethyl O5-Methyl 4-(2-Chlorophenyl)-2-(2-formamidoethoxymethyl)-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate
InChI Formula InChI=1S/C21H25ClN2O6/c1-4-30-21(27)19-16(11-29-10-9-23-12-25)24-13(2)17(20(26)28-3)18(19)14-7-5-6-8-15(14)22/h5-8,12,18,24H,4,9-11H2,1-3H3,(H,23,25)
SMILES CCOC(=O)C1=C(COCCNC=O)NC(=C(C1c2ccccc2Cl)C(=O)OC)C
IUPAC Name 3-O-ethyl 5-O-methyl 4-(2-chlorophenyl)-2-(2-formamidoethoxymethyl)-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate
Molecular Weight (g/mol) 436.886
Formula Weight 436.1401
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.