missing translation for 'onlineSavingsMsg'
Learn More

N-Formyl 10-Oxo Docetaxel, TRC

Product Code. 30395143
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
5 mg
Unit Size:
1mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30395143 5 mg 5mg
30406536 2.5 mg 2.5mg
30404004 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30395143 Supplier Toronto Research Chemicals Supplier No. TRCF7007655MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Formyl 10-Oxo Docetaxel
Analyte or Component Names N-Formyl 10-Oxo Docetaxel
Shipping Condition Room Temperature
Molecular Formula C39H43NO13
Synonym (2aR,4S,4aS,9S,11S,12S,12aR,12bS)-12b-Acetoxy-9-(((2R,3S)-3-formamido-2-hydroxy-3-phenylpropanoyl)oxy)-4,11-dihydroxy-4a,8,13,13-tetramethyl-5,6-dioxo-2a,3,4,4a,5,6,9,10,11,12,12a,12b-dodecahydro-1H-7,11-methanocyclodeca[3,4]benzo[1,2-b]oxet-12-yl Benzoate
InChI Formula InChI=1S/C39H43NO13/c1-20-24(51-35(48)30(45)28(40-19-41)22-12-8-6-9-13-22)17-39(49)33(52-34(47)23-14-10-7-11-15-23)31-37(5,32(46)29(44)27(20)36(39,3)4)25(43)16-26-38(31,18-50-26)53-21(2)42/h6-15,19,24-26,28,30-31,33,43,45,49H,16-18H2,1-5H3,(H,40,41)/t24-,25-,26+,28-,30+,31-,33-,37+,38-,39+/m0/s1
Solution Type Neat
SMILES O=C(C1=C(C)[C@@H](OC([C@H](O)[C@H](C2=CC=CC=C2)NC=O)=O)C[C@]([C@@H](OC(C3=CC=CC=C3)=O)[C@@]4([H])[C@@]5(C)[C@@H](O)C[C@@H]6[C@@]4(OC(C)=O)CO6)(O)C1(C)C)C5=O
Industry Type Pharmaceutical
Molecular Weight (g/mol) 733.76
Quantity 5 mg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.