missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-O-tert-butyl-N-methyl-L-serine, 97%, Thermo Scientific Chemicals

Product Code. 15480399
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15480399 5 g 5g
15490399 250 mg 250mg
15470399 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15480399 Supplier Thermo Scientific Alfa Aesar Supplier No. H63614.06
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 197632-77-2 | Molecular Formula: C23H27NO5 | Molecular Weight: 397.471 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 197632-77-2
Molecular Formula C23H27NO5
Molecular Weight (g/mol) 397.471
MDL Number MFCD02094430
InChI Key PQSAXALAXPNFMG-FQEVSTJZSA-N
Synonym fmoc-n-me-ser tbu-oh, n-fmoc-n-methyl-o-tert-butyl-l-serine, fmoc-n-methyl-o-t-butyl-l-serine, fmoc-n-methyl-o-tert-butyl-l-serine, n-alpha-9-fluorenylmethyloxycarbonyl-n-alpha-methyl-o-t-butyl-l-serine, 2s-3-tert-butoxy-2-9h-fluoren-9-ylmethoxy carbonyl methyl amino propanoic acid, s-2-9h-fluoren-9-yl methoxy carbonyl methyl amino-3-tert-butoxy propanoic acid, fmoc-meser tbu-oh, pubchem19046, fmoc-n-me-serine tbu-oh
PubChem CID 7010370
IUPAC Name (2S)-2-[9H-fluoren-9-ylmethoxycarbonyl(methyl)amino]-3-[(2-methylpropan-2-yl)oxy]propanoic acid
SMILES CC(C)(C)OCC(C(=O)O)N(C)C(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13

Specifications

Melting Point 213°C to 218°C
Color White
Quantity 5 g
Formula Weight 397.46
Percent Purity 97%
Physical Form Crystalline Solid
Chemical Name or Material N-Fmoc-O-tert-butyl-N-methyl-L-serine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.