missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-O-tert-butyl-D-serine, 98%, Thermo Scientific Chemicals

Catalog Number 15438948
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
128107-47-1
C22H25NO5
383.444
MFCD00077071
REITVGIIZHFVGU-LJQANCHMSA-N
fmoc-d-ser tbu-oh, fmoc-o-tert-butyl-d-serine, fmoc-d-ser but-oh, pubchem10040, ksc915q4f, o-tert-butyl-n-fmoc-l-serine, 2r-3-tert-butoxy-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, d-serine, o-1,1-dimethylethyl-n-9h-fluoren-9-ylmethoxy carbonyl, 2r-3-tert-butoxy-2-fluoren-9-ylmethoxy carbonylamino propanoic acid, fmoc-o-tert.butyl-d-serine
7004204
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[(2-methylpropan-2-yl)oxy]propanoic acid
CC(C)(C)OCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
This item is not returnable. View return policy
128107-47-1
C22H25NO5
383.444
MFCD00077071
REITVGIIZHFVGU-LJQANCHMSA-N
fmoc-d-ser tbu-oh, fmoc-o-tert-butyl-d-serine, fmoc-d-ser but-oh, pubchem10040, ksc915q4f, o-tert-butyl-n-fmoc-l-serine, 2r-3-tert-butoxy-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, d-serine, o-1,1-dimethylethyl-n-9h-fluoren-9-ylmethoxy carbonyl, 2r-3-tert-butoxy-2-fluoren-9-ylmethoxy carbonylamino propanoic acid, fmoc-o-tert.butyl-d-serine
7004204
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[(2-methylpropan-2-yl)oxy]propanoic acid
CC(C)(C)OCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 131°C
Quantity 1 g
Beilstein 5309984
Formula Weight 383.44
Percent Purity 98%
Chemical Name or Material N-Fmoc-O-tert-butyl-D-serine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'