missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-O-methyl-D-tyrosine, 98%

Product Code. 15429558
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15429558

Brand: Thermo Scientific Alfa Aesar H63073.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 201335-88-8 | C25H23NO5 | 417.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 201335-88-8
Molecular Formula C25H23NO5
Molecular Weight (g/mol) 417.46
MDL Number MFCD00237393
InChI Key JYQODLWFOPCSCS-UHFFFAOYNA-N
Synonym fmoc-d-tyr me-oh, fmoc-o-methyl-d-tyrosine, fmoc-d-4-methoxyphenylalanine, fmoc-4-methoxy-d-phenylalanine, fmoc-4-methoxy-d-phe-oh, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-methoxyphenyl propanoic acid, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-4-methoxyphenyl propanoic acid, fmoc-d-4-methoxy-phe-oh, fmoc-d-tyr me, fmoc-d-4-meo-phe-oh
PubChem CID 7019819
SMILES COC1=CC=C(CC(NC(=O)OCC2C3=CC=CC=C3C3=CC=CC=C23)C(O)=O)C=C1

Specifications

Color White
IUPAC Name 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(4-methoxyphenyl)propanoic acid
Quantity 250 mg
Formula Weight 417.45
Percent Purity 98%
Physical Form Powder
Chemical Name or Material N-Fmoc-O-methyl-D-tyrosine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.