missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-N-methylglycine, 98%

Product Code. 15459278
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15459278 25 g 25g
15449278 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15459278 Supplier Thermo Scientific Alfa Aesar Supplier No. H62806.14
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 77128-70-2 | C18H19NO5 | 329.352 g/mol

N-Fmoc-N-methylglycine is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Fmoc-N-methylglycine is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Soluble in DMF.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents, heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77128-70-2
Molecular Formula C18H19NO5
Molecular Weight (g/mol) 329.352
MDL Number MFCD00151926
InChI Key CUJSWOOWOONPRH-UHFFFAOYSA-N
Synonym fmoc-sarcosine monohydrate, fmoc-sarcosine hydrate, 2-9h-fluoren-9-yl methoxy carbonyl methyl amino acetic acid hydrate, 9h-fluoren-9-ylmethoxy carbonyl methyl amino acetic acid hydrate, 2-9h-fluoren-9-ylmethoxycarbonyl methyl amino acetic acid;hydrate, fmoc-sarcosinehydrate, ksc915q4d, n-9h-fluoren-9-yl methoxy carbonyl-n-methylglycine-water 1/1
PubChem CID 56763849
IUPAC Name 2-[9H-fluoren-9-ylmethoxycarbonyl(methyl)amino]acetic acid;hydrate
SMILES CN(CC(=O)O)C(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13.O

Specifications

Quantity 25 g
Beilstein 6660958
Solubility Information Soluble in DMF.
Formula Weight 311.33
Percent Purity 98%
Chemical Name or Material N-Fmoc-N-methylglycine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.