missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-N-methyl-L-norleucine, 96%, Thermo Scientific Chemicals

Product Code. 15480999
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15480999

Brand: Thermo Scientific Alfa Aesar H63992.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112883-42-8
Molecular Formula C22H25NO4
Molecular Weight (g/mol) 367.45
MDL Number MFCD00235878
InChI Key RZZXDYZWHUAOEK-GNLPSFAGNA-N
Synonym fmoc-n-me-nle-oh, fmoc-n-methyl-l-norleucine, fmoc-nalpha-methyl-l-norleucine, l-norleucine,n-9h-fluoren-9-ylmethoxy carbonyl-n-methyl, l-norleucine, n-9h-fluoren-9-ylmethoxy carbonyl-n-methyl, 2s-2-9h-fluoren-9-ylmethoxy carbonyl methyl amino hexanoic acid, s-2-9h-fluoren-9-yl methoxy carbonyl methyl amino hexanoic acid, n-alpha-9-fluorenylmethyloxycarbonyl-n-alpha-methyl-l-norleucine, ambotzfaa6460, fmoc-n-me-nle-oh hplc
PubChem CID 7019522
IUPAC Name (2S)-2-[9H-fluoren-9-ylmethoxycarbonyl(methyl)amino]hexanoic acid
SMILES CCCC[C@H](N(C)C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Specifications

Quantity 5 g
Formula Weight 367.44
Percent Purity 96%
Physical Form Solid
Chemical Name or Material N-Fmoc-N-methyl-L-norleucine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.