missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-N-methyl-D-leucine, 97%

Product Code. 15400859
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15400859 1 g 1g
15410859 5 g 5g
15420859 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15400859 Supplier Thermo Scientific Alfa Aesar Supplier No. H63895.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 103478-63-3 | C22H25NO4 | 367.45 g/mol

Tripeptides were prepared by means of coupling of N-Fmoc-N-methyl-D-leucine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tripeptides were prepared by means of coupling of N-Fmoc-N-methyl-D-leucine.

Solubility
Soluble in clear solution (0.3 gram in 2 ml DMF).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103478-63-3
Molecular Formula C22H25NO4
Molecular Weight (g/mol) 367.45
MDL Number MFCD00235877
InChI Key BUJQSIPFDWLNDC-GNLPSFAGNA-N
Synonym fmoc-n-me-d-leu-oh, fmoc-n-methyl-d-leucine, fmoc-d-n-me-leu-oh, d-leucine, n-9h-fluoren-9-ylmethoxy carbonyl-n-methyl, 2r-2-9h-fluoren-9-ylmethoxycarbonyl methyl amino-4-methyl-pentanoic acid, 2r-2-9h-fluoren-9-ylmethoxy carbonyl methyl amino-4-methylpentanoic acid, fmoc-d-meleu-oh, ambotzfaa1517, pubchem18971, h-n-me-d-ala.hcl
PubChem CID 7019520
IUPAC Name (2R)-2-[9H-fluoren-9-ylmethoxycarbonyl(methyl)amino]-4-methylpentanoic acid
SMILES CC(C)C[C@@H](N(C)C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Specifications

Color White
Beilstein 7350417
Solubility Information Soluble in clear solution (0.3 gram in 2 ml DMF).
Quantity 1 g
Formula Weight 367.44
Percent Purity 97%
Physical Form Powder
Chemical Name or Material N-Fmoc-N-methyl-D-leucine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.