missing translation for 'onlineSavingsMsg'
Learn More

N-alpha-FMOC-N-delta-Trityl-L-glutamine, 95%

Product Code. 11975551 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11975551 25 g 25g
11995551 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11975551 Supplier Thermo Scientific Acros Supplier No. 440640250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 132327-80-1 | C39H34N2O5 | 610.71 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 132327-80-1 , 75-09-2
Molecular Formula C39H34N2O5
Molecular Weight (g/mol) 610.71
InChI Key WDGICUODAOGOMO-DHUJRADRSA-N
Synonym fmoc-gln trt-oh, nalpha-fmoc-ndelta-trityl-l-glutamine, fmoc-glutamine trt-oh, n-fmoc-n5-trityl-l-glutamine, n, a-fmoc-n, a-trityl-l-glutamine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-5-oxo-5-tritylamino pentanoic acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-triphenylmethylcarbamoyl butanoic acid, pubchem10017, fmoc-l-gln trt-oh, ksc180c8t
PubChem CID 10919157
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-5-oxo-5-(tritylamino)pentanoic acid
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)NC(=O)CCC(C(=O)O)NC(=O)OCC4C5=CC=CC=C5C6=CC=CC=C46

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 131.0°C to 144.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 25 g
Specific Rotation − 14.20
Formula Weight 610.71
Percent Purity 95%
Physical Form Powder
Chemical Name or Material N-α-FMOC-N-δ-Trityl-L-glutamine
Show More Show Less
compliance-icons
Product Identifier
  • N-alpha-FMOC-N-delta-Trityl-L-glutamine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.