missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-L-proline pentafluorophenyl ester, 98%, Thermo Scientific™

Product Code. 15490159
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15490159

Brand: Thermo Scientific Alfa Aesar H63457.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86060-90-4
Molecular Formula C26H18F5NO4
Molecular Weight (g/mol) 503.425
MDL Number MFCD00065672
InChI Key CQBLOHXKGUNWRV-SFHVURJKSA-N
Synonym fmoc-pro-opfp, fmoc-l-pro-opfp, fmoc-l-proline pentafluorophenyl ester, 1,2-pyrrolidinedicarboxylicacid, 1-9h-fluoren-9-ylmethyl 2-pentafluorophenyl ester, 2s, n-9-fluorenylmethoxycarbonyl-l-proline pentafluorophenyl ester, 1-9h-fluorene-9-ylmethoxycarbonyl-l-proline pentafluorophenyl ester, 1-9h-fluoren-9-ylmethyl 2-2,3,4,5,6-pentafluorophenyl 2s-pyrrolidine-1,2-dicarboxylate, 1-o-9h-fluoren-9-ylmethyl 2-o-2,3,4,5,6-pentafluorophenyl 2s-pyrrolidine-1,2-dicarboxylate
PubChem CID 2733412
IUPAC Name 1-O-(9H-fluoren-9-ylmethyl) 2-O-(2,3,4,5,6-pentafluorophenyl) (2S)-pyrrolidine-1,2-dicarboxylate
SMILES C1CC(N(C1)C(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24)C(=O)OC5=C(C(=C(C(=C5F)F)F)F)F

Specifications

Melting Point 127°C to 129°C
Beilstein 4731096
Quantity 5 g
Formula Weight 503.42
Percent Purity 98%
Physical Form Solid
Chemical Name or Material N-Fmoc-L-proline pentafluorophenyl ester
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.