missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-L-norleucine, 98%, Thermo Scientific Chemicals

Product Code. 11349769
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11349769 25 g 25g
11329769 1 g 1g
11339769 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11349769 Supplier Thermo Scientific Alfa Aesar Supplier No. B22475.14
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77284-32-3
Molecular Formula C21H23NO4
Molecular Weight (g/mol) 353.42
MDL Number MFCD00037537
InChI Key VCFCFPNRQDANPN-IBGZPJMESA-N
Synonym fmoc-nle-oh, fmoc-l-norleucine, n-fmoc-l-norleucine, s-2-fmoc-amino caproic acid, s-2-9h-fluoren-9-yl methoxy carbonyl amino hexanoic acid, s-2-9h-fluoren-9-ylmethoxycarbonylamino-hexanoic acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid, fmoc-norleucine, fmocnleoh, fmoc-nle
PubChem CID 7009636
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)hexanoic acid
SMILES CCCC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Specifications

Melting Point 140°C to 142°C
Quantity 25 g
Formula Weight 353.42
Percent Purity 98%
Chemical Name or Material N-Fmoc-L-norleucine
compliance-icons
Product Identifier
  • N-Fmoc-L-norleucine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: N-Fmoc-L-norleucine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.