missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-L-norleucine, 98%, Thermo Scientific Chemicals

Product Code. 11329769
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11329769 1 g 1g
11339769 5 g 5g
11349769 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11329769 Supplier Thermo Scientific Alfa Aesar Supplier No. B22475.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77284-32-3
Molecular Formula C21H23NO4
Molecular Weight (g/mol) 353.42
MDL Number MFCD00037537
InChI Key VCFCFPNRQDANPN-IBGZPJMESA-N
Synonym fmoc-nle-oh, fmoc-l-norleucine, n-fmoc-l-norleucine, s-2-fmoc-amino caproic acid, s-2-9h-fluoren-9-yl methoxy carbonyl amino hexanoic acid, s-2-9h-fluoren-9-ylmethoxycarbonylamino-hexanoic acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid, fmoc-norleucine, fmocnleoh, fmoc-nle
PubChem CID 7009636
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)hexanoic acid
SMILES CCCC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Specifications

Melting Point 140°C to 142°C
Quantity 1 g
Formula Weight 353.42
Percent Purity 98%
Chemical Name or Material N-Fmoc-L-norleucine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.