missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-L-methionine sulfoxide, 98%

Product Code. 15439888
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15439888

Brand: Thermo Scientific Alfa Aesar H63271.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 76265-70-8 | C20H21NO5S | 387.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76265-70-8
Molecular Formula C20H21NO5S
Molecular Weight (g/mol) 387.45
MDL Number MFCD00077064
InChI Key CEHRSUBRZOGRSW-HSYKDVHTSA-N
Synonym fmoc-met o-oh, fmoc-l-methionine sulfoxide, 2s-2-9h-fluoren-9-yl methoxy carbonyl amino-4-methylsulfinyl butanoic acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-methanesulfinylbutanoic acid, ambotzfaa1405, n-9-fluorenylmethoxycarbonyl-methionine sulfoxide, n-alpha-9-fluorenylmethyloxycarbonyl-l-methioninesulfoxid
PubChem CID 7408215
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-methylsulfinylbutanoic acid
SMILES CS(=O)CC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Specifications

Color White
Quantity 5 g
Formula Weight 387.45
Percent Purity 98%
Physical Form Powder
Chemical Name or Material N-Fmoc-L-methionine sulfoxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.