missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-L-glutamic acid, 95%

Product Code. 15489378
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15489378 5 g 5g
15499378 25 g 25g
15409388 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15489378 Supplier Thermo Scientific Alfa Aesar Supplier No. H62936.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 121343-82-6 | C20H19NO6 | 369.373 g/mol

N-Fmoc-L-glutamic acid nonessential amino acid that is important to the mammalian central nervous system. It is also a neurotransmitter for cone photoreceptors in the human brain and is used as a treatment for patients who have liver disease accompanied by encephalopathy (a condition termed ‘hepatic coma’).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121343-82-6
Molecular Formula C20H19NO6
Molecular Weight (g/mol) 369.373
MDL Number MFCD00237657
InChI Key QEPWHIXHJNNGLU-KRWDZBQOSA-N
Synonym fmoc-glu-oh, fmoc-l-glutamic acid, s-2-9h-fluoren-9-yl methoxy carbonyl amino pentanedioic acid, n-fmoc-l-glutamic acid, fmoc-glu, n-9h-fluoren-9-ylmethoxy carbonyl-l-glutamic acid, l-glutamic acid, n-9h-fluoren-9-ylmethoxy carbonyl, 2s-2-fluoren-9-ylmethoxy carbonylamino pentanedioic acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino pentanedioic acid
PubChem CID 7019018
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)pentanedioic acid
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CCC(=O)O)C(=O)O

Specifications

Quantity 5 g
Beilstein 3633624
Solubility Information Slightly soluble in water.
Formula Weight 369.37
Percent Purity 95%
Chemical Name or Material N-Fmoc-L-glutamic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.