missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-L-aspartic acid, 95%

Product Code. 15468818
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15468818 5 g 5g
15478818 25 g 25g
15488818 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15468818 Supplier Thermo Scientific Alfa Aesar Supplier No. H62274.06
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 119062-05-4 | C19H17NO6 | 355.346 g/mol

N-Fmoc-L-aspartic acid is used to biosynthesize other amino acids with in the human body.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Fmoc-L-aspartic acid is used to biosynthesize other amino acids with in the human body.

Solubility
Soluble in methanol, and water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 119062-05-4
Molecular Formula C19H17NO6
Molecular Weight (g/mol) 355.346
MDL Number MFCD00237654
InChI Key KSDTXRUIZMTBNV-INIZCTEOSA-N
Synonym fmoc-asp-oh, fmoc-l-aspartic acid, n-fmoc-l-aspartic acid, fmoc-l-asp-oh, fmoc-aspartic acid, n-9h-fluoren-9-ylmethoxy carbonyl-l-aspartic acid, s-2-9h-fluoren-9-yl methoxy carbonylamino succinic acid, s-2-9h-fluoren-9-yl methoxy carbonyl amino succinic acid, aspartic acid, n-9h-fluoren-9-ylmethoxy carbonyl, n-9-fluorenylmethoxycarbonyl-l-aspartic acid
PubChem CID 7019016
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)butanedioic acid
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC(=O)O)C(=O)O

Specifications

Quantity 5 g
Beilstein 3632557
Solubility Information Soluble in methanol,and water.
Formula Weight 355.34
Percent Purity 95%
Chemical Name or Material N-Fmoc-L-aspartic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.