missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-L-aspartic acid 4-tert-butyl ester, 98%

Product Code. 15455168
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 15455168

Brand: Thermo Scientific Alfa Aesar H59007.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 71989-14-5 | C23H25NO6 | 411.454 g/mol

L-Aspartic acid is a nonessential amino acid that is used to biosynthesize other amino acids within the human body.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Aspartic acid is a nonessential amino acid that is used to biosynthesize other amino acids within the human body.

Solubility
Slightly soluble in water.

Notes
Refrigerate. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71989-14-5
Molecular Formula C23H25NO6
Molecular Weight (g/mol) 411.454
MDL Number MFCD00037131
InChI Key FODJWPHPWBKDON-IBGZPJMESA-N
Synonym fmoc-asp otbu-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-4-tert-butoxy-4-oxobutanoic acid, fmoc-asp obut, fmoc-l-aspartic acid beta-tert-butyl ester, fmoc-asp ot-bu-oh, fmoc-l-asp tbu, fmocasp otbu-oh, fmoc-asp obu-oh, fmoc-asp tbu-oh, fmoc-l-aspartic acid 4-tert-butyl ester
PubChem CID 2724635
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-[(2-methylpropan-2-yl)oxy]-4-oxobutanoic acid
SMILES CC(C)(C)OC(=O)CC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13

Specifications

Melting Point 148°C to 150°C (decomposition)
Quantity 5 g
Beilstein 3635671
Solubility Information Slightly soluble in water.
Optical Rotation −24° (c=1 in DMF)
Formula Weight 411.45
Percent Purity 98%
Chemical Name or Material N-Fmoc-L-aspartic acid 4-tert-butyl ester

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.