missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-L-aspartic acid 4-benzyl ester, 97%

Product Code. 15488608
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 15488608

Brand: Thermo Scientific Alfa Aesar H62011.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 86060-84-6 | C26H23NO6 | 445.47 g/mol

Used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as pharmaceutical intermediates.

Solubility
Soluble in methanol.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86060-84-6
Molecular Formula C26H23NO6
Molecular Weight (g/mol) 445.47
MDL Number MFCD00065630
InChI Key OQGAELAJEGGNKG-UHFFFAOYNA-N
Synonym fmoc-asp obzl-oh, fmoc-l-aspartic acid 4-benzyl ester, l-fmoc-aspartic acid beta-benzyl ester, fmoc-l-asp obzl-oh, 2s-4-benzyloxy-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-oxobutanoic acid, fmoc-l-aspartic acid beta-benzyl ester, s-2-9h-fluoren-9-yl methoxy carbonyl amino-4-benzyloxy-4-oxobutanoic acid, n-fmoc-l-aspartic acid beta-benzyl ester, fmoc-l-asp obn, fmoc-l-asp bzl-oh
PubChem CID 13966931
SMILES OC(=O)C(CC(=O)OCC1=CC=CC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Melting Point 120°C to 130°C
Quantity 25 g
Beilstein 4609615
Solubility Information Soluble in methanol.
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-oxo-4-phenylmethoxybutanoic acid
Formula Weight 445.46
Percent Purity 97%
Chemical Name or Material N-Fmoc-L-aspartic acid 4-benzyl ester

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.