missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-L-alpha-phenylglycine, 98%

Product Code. 15459448
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 15459448

Brand: Thermo Scientific Alfa Aesar H63017.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 102410-65-1 | C23H19NO4 | 373.408 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102410-65-1
Molecular Formula C23H19NO4
Molecular Weight (g/mol) 373.408
MDL Number MFCD00155632
InChI Key PCJHOCNJLMFYCV-NRFANRHFSA-N
Synonym fmoc-phg-oh, fmoc-l-phenylglycine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-2-phenylacetic acid, n-fmoc-l-2-phenylglycine, fmoc-s-phenylglycine, n-9h-fluoren-9-ylmethoxy carbonyl-l-2-phenylglycine, n-alpha-9-fluorenylmethyloxycarbonyl-l-phenylglycine, s-9h-fluoren-9-ylmethoxycarbonylamino-phenyl-acetic acid, s-9h-fluoren-9-ylmethoxy carbonyl amino phenyl acetic acid, 2s-9h-fluoren-9-yl-methoxy carbonyl amino phenyl acetic acid
PubChem CID 7269367
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-2-phenylacetic acid
SMILES C1=CC=C(C=C1)C(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24

Specifications

Beilstein 5309005
Quantity 5 g
Formula Weight 373.4
Percent Purity 98%
Physical Form Solid
Chemical Name or Material N-Fmoc-L-alpha-phenylglycine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.