missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-L-alanine, 95%

Product Code. 15425768
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15425768 25 g 25g
15415768 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15425768 Supplier Thermo Scientific Alfa Aesar Supplier No. H59438.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 35661-39-3 | C18H17NO4 | 311.337 g/mol

N-Fmoc-L-alanine is potentially useful for proteomics studies and solid phase peptide synthesis techniques. Alanine is one of the simplest amino acids - a methyl group as the side chain. This small side chain confers a high degree of flexibility when incorporated into a polypeptide chain. The Fmoc group is typically removed with a base such as pyridine - an orthogonal de-protection strategy to the acid labilie Boc group.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Fmoc-L-alanine is potentially useful for proteomics studies and solid phase peptide synthesis techniques. Alanine is one of the simplest amino acids - a methyl group as the side chain. This small side chain confers a high degree of flexibility when incorporated into a polypeptide chain. The Fmoc group is typically removed with a base such as pyridine - an orthogonal de-protection strategy to the acid labilie Boc group.

Solubility
Soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35661-39-3
Molecular Formula C18H17NO4
Molecular Weight (g/mol) 311.337
MDL Number MFCD00037139
InChI Key QWXZOFZKSQXPDC-NSHDSACASA-N
Synonym fmoc-ala-oh, fmoc-l-alanine, fmoc-l-alpha-alanine, 9-fmoc-l-alanine, fmoc-l-ala-oh, n-fmoc-l-alanine, n-9h-fluoren-9-ylmethoxy carbonyl-l-alanine, n-9-fluorenylmethoxycarbonyl-l-alanine, fmoc-alanine, fmoc-l-ala
PubChem CID 6364642
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid
SMILES CC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13

Specifications

Melting Point 147°C to 153°C
Quantity 25 g
Beilstein 2225975
Solubility Information Soluble in water.
Optical Rotation −18° (c=1 in DMF)
Formula Weight 311.34
Percent Purity 95%
Chemical Name or Material N-Fmoc-L-alanine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.