missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-D-valine, 98%

Product Code. p-7040005
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15455938 1 g 1g
15465938 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15455938 Supplier Thermo Scientific Alfa Aesar Supplier No. H59570.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 84624-17-9 | C20H21NO4 | 339.39 g/mol

It is potentially useful for proteomics studies and solid phase peptide synthesis techniques. Valine is one of the simplest amino acids - an isopropyl group as the side chain. This smaller side chain confers a fairly high degree of flexibility when incorporated into a polypeptide chain. The Fmoc group is typically removed with a base such as pyridine - an orthogonal de-protection strategy to the acid labilie Boc group.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is potentially useful for proteomics studies and solid phase peptide synthesis techniques. Valine is one of the simplest amino acids - an isopropyl group as the side chain. This smaller side chain confers a fairly high degree of flexibility when incorporated into a polypeptide chain. The Fmoc group is typically removed with a base such as pyridine - an orthogonal de-protection strategy to the acid labilie Boc group.

Solubility
Solubility in methanol (almost transparency). Slightly soluble in water.

Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84624-17-9
Molecular Formula C20H21NO4
Molecular Weight (g/mol) 339.39
MDL Number MFCD00062953
InChI Key UGNIYGNGCNXHTR-GOSISDBHSA-N
Synonym fmoc-d-val-oh, fmoc-d-valine, n-fmoc-d-valine, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-methylbutanoic acid, n-9h-fluoren-9-ylmethoxy carbonyl-d-valine, n-9-fluorenylmethoxycarbonyl-d-valine, 9-fluorenylmethoxycarbonyl-d-valine, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-methylbutanoic acid, n-9-fluorenylmethyloxycarbonyl-d-valine, 9h-fluoren-9-yl methoxy carbonyl-d-valine
PubChem CID 1549479
SMILES CC(C)[C@@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Specifications

Melting Point 143°C to 144°C
Quantity 1 g
Beilstein 6489548
IUPAC Name (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-methylbutanoic acid
Formula Weight 339.39
Percent Purity 98%
Chemical Name or Material N-Fmoc-D-valine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.