missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-D-allo-isoleucine, 98%, Thermo Scientific Chemicals

Catalog Number p-7054988
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
118904-37-3
C21H23NO4
353.42
MFCD00077062
QXVFEIPAZSXRGM-KINVLMLONA-N
fmoc-d-allo-ile-oh, fmoc-d-allo-isoleucine, fmoc-d-allo-lle-oh, 2r,3s-n-fmoc-2-amino-3-methylpentanoic acid, 2r,3s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-methylpentanoic acid, d-alloisoleucine, n-9h-fluoren-9-ylmethoxy carbonyl, 2r,3s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-methylpentanoic acid, ambotzfaa1328, pubchem15619
6992517
(2R,3S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-methylpentanoic acid
CC[C@H](C)[C@@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
This item is not returnable. View return policy
118904-37-3
C21H23NO4
353.42
MFCD00077062
QXVFEIPAZSXRGM-KINVLMLONA-N
fmoc-d-allo-ile-oh, fmoc-d-allo-isoleucine, fmoc-d-allo-lle-oh, 2r,3s-n-fmoc-2-amino-3-methylpentanoic acid, 2r,3s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-methylpentanoic acid, d-alloisoleucine, n-9h-fluoren-9-ylmethoxy carbonyl, 2r,3s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-methylpentanoic acid, ambotzfaa1328, pubchem15619
6992517
(2R,3S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-methylpentanoic acid
CC[C@H](C)[C@@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

N-Fmoc-D-allo-isoleucine used as a intermediate in pharmaceutical and chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Fmoc-D-allo-isoleucine used as a intermediate in pharmaceutical and chemical synthesis.

Solubility
Slightly soluble in water.

Notes
Refrigerate at -20°C. Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Solubility Information Slightly soluble in water.
Formula Weight 353.41
Percent Purity 98%
Quantity 250 mg
Physical Form Solid
Chemical Name or Material N-Fmoc-D-allo-isoleucine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'