missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-4-methyl-L-phenylalanine, 95%

Product Code. 15468597
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15468597 1 g 1g
15488597 250 mg 250mg
15478597 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15468597 Supplier Thermo Scientific Alfa Aesar Supplier No. H52073.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 199006-54-7 | C25H23NO4 | 401.462 g/mol

N-Fmoc-4-methyl-L-phenylalanine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Fmoc-4-methyl-L-phenylalanine is used as pharmaceutical intermediate.

Solubility
Soluble in Dimethylformamide(DMF).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 199006-54-7
Molecular Formula C25H23NO4
Molecular Weight (g/mol) 401.462
MDL Number MFCD00270197
InChI Key UXLHLZHGQPDMJQ-QHCPKHFHSA-N
Synonym fmoc-phe 4-me-oh, fmoc-l-4-methylphe, fmoc-4-methyl-l-phenylalanine, fmoc-l-4-methylphenylalanine, fmoc-4-methy-l-phenylalanine, fmoc-l-phe 4-me-oh, fmoc-p-me-phe-oh, n-fmoc-4-methyl-l-phenylalanine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-p-tolyl propanoic acid, phenylalanine, n-9h-fluoren-9-ylmethoxy carbonyl-4-methyl
PubChem CID 7129826
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(4-methylphenyl)propanoic acid
SMILES CC1=CC=C(C=C1)CC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24

Specifications

Quantity 1 g
Beilstein 8799411
Solubility Information Soluble in Dimethylformamide(DMF).
Formula Weight 401.46
Percent Purity 95%
Chemical Name or Material N-Fmoc-4-methyl-L-phenylalanine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.