missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-4-chloro-D-phenylalanine, 98%, Thermo Scientific™

Product Code. 15400369
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142994-19-2
Molecular Formula C24H20ClNO4
Molecular Weight (g/mol) 421.88
MDL Number MFCD00077066
InChI Key CQPNKLNINBUUOM-UHFFFAOYNA-N
Synonym fmoc-4-chloro-d-phenylalanine, fmoc-d-phe 4-cl-oh, fmoc-d-4-chlorophe, fmoc-d-4-chlorophenylalanine, fmoc-d-pclphe-oh, fmoc-p-chloro-d-phe-oh, fmoc-4-chloro-d-phe-oh, fmoc-pcl-d-phe-oh, d-phenylalanine, 4-chloro-n-9h-fluoren-9-ylmethoxy carbonyl, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-chlorophenyl propanoic acid
PubChem CID 16213163
SMILES OC(=O)C(CC1=CC=C(Cl)C=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Color White
Beilstein 8726004
IUPAC Name 3-(4-chlorophenyl)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid
Formula Weight 421.87
Percent Purity 98%
Quantity 1 g
Physical Form Powder
Chemical Name or Material N-Fmoc-4-chloro-D-phenylalanine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'