missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-3-cyclohexyl-D-alanine, 96%, Thermo Scientific™

Catalog Number 15418908
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
144701-25-7
C24H27NO4
393.48
MFCD00151912
HIJAUEZBPWTKIV-UHFFFAOYNA-N
fmoc-d-cha-oh, fmoc-3-cyclohexyl-d-alanine, fmoc-beta-cyclohexyl-d-alanine, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-cyclohexylpropanoic acid, n-alpha-fmoc-beta-cyclohexyl-d-alanine, n-alpha-9-fluorenylmethyloxycarbonyl-d-cyclohexylalanine, 2r-3-cyclohexyl-2-9h-fluoren-9-ylmethoxycarbonylamino propanoic acid, 2r-3-cyclohexyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, rarechem em wb 0095, fmoc-beta-cyclohexyl-d-ala-oh
978327
OC(=O)C(CC1CCCCC1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
This item is not returnable. View return policy
144701-25-7
C24H27NO4
393.48
MFCD00151912
HIJAUEZBPWTKIV-UHFFFAOYNA-N
fmoc-d-cha-oh, fmoc-3-cyclohexyl-d-alanine, fmoc-beta-cyclohexyl-d-alanine, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-cyclohexylpropanoic acid, n-alpha-fmoc-beta-cyclohexyl-d-alanine, n-alpha-9-fluorenylmethyloxycarbonyl-d-cyclohexylalanine, 2r-3-cyclohexyl-2-9h-fluoren-9-ylmethoxycarbonylamino propanoic acid, 2r-3-cyclohexyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, rarechem em wb 0095, fmoc-beta-cyclohexyl-d-ala-oh
978327
OC(=O)C(CC1CCCCC1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
N-Fmoc-3-cyclohexyl-D-alanine is used as fine chemical intermediate.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Color White
Beilstein 8234680
IUPAC Name (2R)-3-cyclohexyl-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid
Formula Weight 393.48
Percent Purity 96%
Quantity 1 g
Physical Form Powder
Chemical Name or Material N-Fmoc-3-cyclohexyl-D-alanine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'