missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-3-(4-pyridyl)-D-alanine, 95%

Product Code. 15498757
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15498757 250 mg 250mg
15478757 1 g 1g
15488757 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15498757 Supplier Thermo Scientific Alfa Aesar Supplier No. H52193.MD
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 205528-30-9 | C23H20N2O4 | 388.42 g/mol

N-Fmoc-3-(4-pyridyl)-D-alanine is an Fmoc protected alanine derivative that is potentially useful for proteomics studies and solid phase peptide synthesis techniques.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Fmoc-3-(4-pyridyl)-D-alanine is an Fmoc protected alanine derivative that is potentially useful for proteomics studies and solid phase peptide synthesis techniques.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 205528-30-9
Molecular Formula C23H20N2O4
Molecular Weight (g/mol) 388.42
MDL Number MFCD00672567
InChI Key SCSSXJVRZMQUKA-UHFFFAOYNA-N
Synonym fmoc-d-4-pyridylalanine, fmoc-d-4-pal-oh, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-pyridin-4-yl propanoic acid, fmoc-beta-4-pyridyl-d-ala-oh, n-fmoc-3-4-pyridyl-d-alanine, fmoc-4'-pyridyl-d-ala, fmoc-3-4-pyridyl-d-alanine, fmoc-d-3-4-pyridyl-alanine, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-pyridin-4-yl propanoic acid, r-2-9h-fluoren-9-ylmethoxycarbonylamino-3-pyridin-4-yl-propionic acid
PubChem CID 978323
IUPAC Name (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-pyridin-4-ylpropanoic acid
SMILES OC(=O)C(CC1=CC=NC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Quantity 250 mg
Formula Weight 388.42
Percent Purity 95%
Chemical Name or Material N-Fmoc-3-(4-pyridyl)-D-alanine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.