missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-3,3-diphenyl-L-alanine, 95%

Product Code. 15458727
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15458727 250 mg 250mg
15438727 1 g 1g
15448727 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15458727 Supplier Thermo Scientific Alfa Aesar Supplier No. H52176.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 201484-50-6 | C30H25NO4 | 463.533 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 201484-50-6
Molecular Formula C30H25NO4
Molecular Weight (g/mol) 463.533
MDL Number MFCD00672337
InChI Key PENQOTJCVODUQU-NDEPHWFRSA-N
Synonym fmoc-l-3,3-diphenylalanine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3,3-diphenylpropanoic acid, fmoc-3,3-diphenyl-l-alanine, fmoc-beta-phenyl-phe-oh, 2s-2-9h-fluoren-9-ylmethoxycarbonylamino-3,3-diphenyl-propanoic acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3,3-diphenylpropanoic acid, fmoc-3,3-diphenyl-d-alaine, fmoc-3,3-diphenyl-l-alaine, fmoc-,-diphenyl-ala-oh, fmoc-ala 3,3-diphenyl-oh
PubChem CID 2761488
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3,3-diphenylpropanoic acid
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)C(C(=O)O)NC(=O)OCC3C4=CC=CC=C4C5=CC=CC=C35

Specifications

Melting Point 125°C to 129°C
Quantity 250 mg
UN Number UN3077
Beilstein 8873562
Formula Weight 463.53
Percent Purity 95%
Chemical Name or Material N-Fmoc-3,3-diphenyl-L-alanine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.