missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-2-octyl-L-glycine, 95%

Product Code. 15422689
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15422689 1 g 1g
15442689 250 mg 250mg
15432689 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15422689 Supplier Thermo Scientific Alfa Aesar Supplier No. H66031.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 193885-59-5 | C25H31NO4 | 409.526 g/mol

A homolog of lipophilic building block for SAR studies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A homolog of lipophilic building block for SAR studies.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 193885-59-5
Molecular Formula C25H31NO4
Molecular Weight (g/mol) 409.526
MDL Number MFCD01311753
InChI Key LSMLSLHVRMSYPT-QHCPKHFHSA-N
Synonym fmoc-octyl-gly-oh, fmoc-2s-2-amino-decanoic acid, s-2-9h-fluoren-9-yl methoxy carbonyl amino decanoic acid, ambotzfaa1598, fmoc-s-octylglycine, s-n-fmoc-octylglycine-oh, n-alpha-9-fluorenylmethyloxycarbonyl-l-octylglycine, 2s-2-9h-fluoren-9-ylmethoxycarbonylamino decanoic acid, s-2-9h-fluoren-9-yl methoxy carbonylamino decanoic acid, 2s-2-9h-fluoren-9-yl methoxy carbonyl amino decanoic acid
PubChem CID 23257324
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)decanoic acid
SMILES CCCCCCCCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13

Specifications

Melting Point 140°C to 143°C
Color White
Solubility Information Slightly soluble in water.
Quantity 1 g
Formula Weight 409.53
Percent Purity 98%
Physical Form Powder
Chemical Name or Material N-Fmoc-2-octyl-L-glycine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.