missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-1-trityl-D-histidine, 98%

Product Code. 15449028
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 15449028

Brand: Thermo Scientific Alfa Aesar H62504.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 135610-90-1 | C40H33N3O4 | 619.721 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 135610-90-1
Molecular Formula C40H33N3O4
Molecular Weight (g/mol) 619.721
MDL Number MFCD00077061
InChI Key XXMYDXUIZKNHDT-DIPNUNPCSA-N
Synonym fmoc-d-his trt-oh, n-fmoc-n'-trityl-d-histidine, d-histidine, n-9h-fluoren-9-ylmethoxy carbonyl-1-triphenylmethyl, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-1-triphenylmethyl imidazol-4-yl propanoic acid, pubchem18930, fmoc-d-his 1-trt-oh, fmoc-d-his-trt-oh, n-fmoc-1-trityl-d-histidine, fmoc-n-im-trityl-d-histidine, nalpha-fmoc-nim-trityl-d-histidine
PubChem CID 44828577
IUPAC Name (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(1-tritylimidazol-4-yl)propanoic acid
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)N4C=C(N=C4)CC(C(=O)O)NC(=O)OCC5C6=CC=CC=C6C7=CC=CC=C57

Specifications

Quantity 5 g
Optical Rotation −80° (c=1 in Chloroform)
Formula Weight 619.71
Percent Purity 98%
Chemical Name or Material N-Fmoc-1-trityl-D-histidine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.