missing translation for 'onlineSavingsMsg'
Learn More

N-Fluorobenzenesulfonimide, TRC

Product Code. 30393355
Change view
Click to view available options
Quantity:
500 mg
1 g
2.5 g
Unit Size:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30393355 500 mg -
30404821 1 g -
30403741 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30393355 Supplier Toronto Research Chemicals Supplier No. TRCF596833500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 133745-75-2
Molecular Formula C12H10FNO4S2
Molecular Weight (g/mol) 315.34
IUPAC Name N-(benzenesulfonyl)-N-fluorobenzenesulfonamide
SMILES FN(S(=O)(=O)c1ccccc1)S(=O)(=O)c2ccccc2

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C12H10FNO4S2/c13-14(19(15,16)11-7-3-1-4-8-11)20(17,18)12-9-5-2-6-10-12/h1-10H
Formula Weight 315.0035
Chemical Name or Material N-Fluorobenzenesulfonimide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.