missing translation for 'onlineSavingsMsg'
Learn More

N-Feruloyl Serotonin, TRC

Product Code. 30398344
Change view
Click to view available options
Quantity:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30398344 250 mg
30392390 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30398344 Supplier Toronto Research Chemicals Supplier No. TRCF309000250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68573-23-9
Molecular Formula C20H20N2O4
Molecular Weight (g/mol) 352.38
Synonym N-[2-(5-Hydroxy-1H-indol-3-yl)ethyl]-3-(4-hydroxy-3-methoxyphenyl)-2-propenamide, N-Feruloylserotonin, NSC 369502
IUPAC Name (E)-N-(2-(5-hydroxy-1H-indol-3-yl)ethyl)-3-(4-hydroxy-3-methoxyphenyl)acrylamide
SMILES OC1=CC=C(NC=C2CCNC(/C=C/C3=CC(OC)=C(O)C=C3)=O)C2=C1

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C20H20N2O4/c1-26-19-10-13(2-6-18(19)24)3-7-20(25)21-9-8-14-12-22-17-5-4-15(23)11-16(14)17/h2-7,10-12,22-24H,8-9H2,1H3,(H,21,25)/b7-3+
Formula Weight 352.14
Percent Purity >95%
Chemical Name or Material N-Feruloyl Serotonin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.