missing translation for 'onlineSavingsMsg'
Learn More

N-Ethylmethylamine, TRC

Product Code. 30136985
Change view
Click to view available options
Quantity:
1 g
5 g
50 g
Unit Size:
1g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136985 50 g 50g
30135789 5 g 5g
30010098 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136985 Supplier Toronto Research Chemicals Supplier No. TRCE92485050G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 624-78-2
Molecular Formula C3 H9 N
Molecular Weight (g/mol) 59.11
Synonym Ethanamine, N-methyl- (9CI, ACI), Ethylamine, N-methyl- (7CI, 8CI), N-Methylethanamine (ACI), Ethylmethylamine, Methylaminoethane, Methylethylamine, N-Ethyl-N-methylamine, N-Ethylmethylamine, N-Methyl-N-ethylamine, N-Methylethylamine, NMEA
IUPAC Name N-methylethanamine
SMILES CCNC

Specifications

Quantity 50 g
InChI Formula InChI=1S/C3H9N/c1-3-4-2/h4H,3H2,1-2H3
Formula Weight 59.0735
Chemical Name or Material N-Ethylmethylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.