missing translation for 'onlineSavingsMsg'
Learn More

N-Ethylmaleimide, 99+%, Thermo Scientific Chemicals

Catalog Number 11368518
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
128-53-0
C6H7NO2
125.127
MFCD00005509
HDFGOPSGAURCEO-UHFFFAOYSA-N
n-ethylmaleimide, ethylmaleimide, 1-ethyl-1h-pyrrole-2,5-dione, maleimide, n-ethyl, 1h-pyrrole-2,5-dione, 1-ethyl, maleic acid n-ethylimide, nem, usaf b-121, unii-o3c74acm9v, maleic acid-n-ethylimide
4362
CHEBI:44485
1-ethylpyrrole-2,5-dione
CCN1C(=O)C=CC1=O
This item is not returnable. View return policy
128-53-0
C6H7NO2
125.127
MFCD00005509
HDFGOPSGAURCEO-UHFFFAOYSA-N
n-ethylmaleimide, ethylmaleimide, 1-ethyl-1h-pyrrole-2,5-dione, maleimide, n-ethyl, 1h-pyrrole-2,5-dione, 1-ethyl, maleic acid n-ethylimide, nem, usaf b-121, unii-o3c74acm9v, maleic acid-n-ethylimide
4362
CHEBI:44485
1-ethylpyrrole-2,5-dione
CCN1C(=O)C=CC1=O

N-Ethylmaleimide is a protein thiol modifier which inhibits apoptotic DNA fragmentation. It is a sulfhydryl reagent and finds application in experimental biochemical studies as well as in enzymology. It is involved in the modification of cysteine residues in proteins and peptides. It acts as a Michael acceptor and reacts with nucelophiles like thiols. It is employed in the inhibition of Mg2+ dependent internucleosomal DNA fragmentation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 43°C to 46°C
Boiling Point 209°C to 211°C
Flash Point 73°C (163°F)
Assay Percent Range >99+%
Quantity 1 g
Beilstein 112448
Sensitivity Light sensitive
Solubility Information Soluble in ethanol,water,chloroform,methanol and toluene.
Formula Weight 125.13
Physical Form Crystalline
Chemical Name or Material N-Ethylmaleimide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'