missing translation for 'onlineSavingsMsg'
Learn More

N-Ethylmaleimide, 98+%

Product Code. 10112970 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
250 g
Unit Size:
10g
250g
50g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10112970 50 g 50g
10266200 5 g 5g
10021790 10 g 10g
11945581 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10112970 Supplier Thermo Scientific Acros Supplier No. 156100500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 128-53-0 | C6H7NO2 | 125.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 128-53-0
Molecular Formula C6H7NO2
Molecular Weight (g/mol) 125.13
InChI Key HDFGOPSGAURCEO-UHFFFAOYSA-N
Synonym n-ethylmaleimide, ethylmaleimide, 1-ethyl-1h-pyrrole-2,5-dione, maleimide, n-ethyl, 1h-pyrrole-2,5-dione, 1-ethyl, maleic acid n-ethylimide, nem, usaf b-121, unii-o3c74acm9v, maleic acid-n-ethylimide
PubChem CID 4362
ChEBI CHEBI:44485
IUPAC Name 1-ethylpyrrole-2,5-dione
SMILES CCN1C(=O)C=CC1=O

Specifications

Melting Point 41.0°C to 45.0°C
Color White
Boiling Point 210.0°C
Flash Point 73°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Quantity 50 g
Beilstein 21,IV,4629
Merck Index 15,3877
Solubility Information Solubility in water: soluble. Other solubilities: 1115g/L toluene (20°C)
Formula Weight 125.13
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material N-Ethylmaleimide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • N-Ethylmaleimide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
Hazard Statement
  • H300-Fatal if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Maleic acid N-ethylimide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.