missing translation for 'onlineSavingsMsg'
Learn More

N-Ethylisopropylamine, 98%

Product Code. 11346786
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11346786 1 g 1g
11356786 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11346786 Supplier Thermo Scientific Alfa Aesar Supplier No. L04089.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 19961-27-4 | C5H13N | 87.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19961-27-4
Molecular Formula C5H13N
Molecular Weight (g/mol) 87.17
MDL Number MFCD00015105
InChI Key RIVIDPPYRINTTH-UHFFFAOYSA-N
Synonym n-ethylisopropylamine, ethylisopropylamine, 2-propanamine, n-ethyl, ethyl propan-2-yl amine, diethylamine, 1-methyl, isopropylamine, n-ethyl, n-ethyl-2-propanamine, n-ethyl-n-isopropylamine, isopropylethylamine, ethyl isopropylamine
PubChem CID 88318
SMILES CCNC(C)C

Specifications

Density 0.72
Boiling Point 71°C to 73°C
Flash Point 16°C (60°F)
Refractive Index 1.392
Quantity 1 g
UN Number UN2733
Beilstein 505952
IUPAC Name ethyl(propan-2-yl)amine
Formula Weight 87.17
Percent Purity 98%
Chemical Name or Material N-Ethylisopropylamine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • N-Ethylisopropylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: N-Ethylisopropylamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.