missing translation for 'onlineSavingsMsg'
Learn More

N-Ethyldicyclohexylamine, 96%

Product Code. 10490921 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10490921 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10490921 Supplier Thermo Scientific Acros Supplier No. 314730050
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 7175-49-7 | C14H27N | 209.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7175-49-7
CAS Min % 95.0
CAS Max % 100.0
Color Colorless
Density 0.9000g/mL
Boiling Point 114°C to 115°C (3.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Molecular Formula C14H27N
Linear Formula (C6H11)2NC2H5
Refractive Index 1.4840 to 1.487
MDL Number MFCD00003846
Quantity 5 g
Specific Gravity 0.9
Synonym n-ethyldicyclohexylamine, ethyldicyclohexylamine, cyclohexanamine, n-cyclohexyl-n-ethyl, dicyclohexylethylamine, dicyclohexylamine, n-ethyl, n-cyclohexyl-n-ethylcyclohexylamine, ethyl-dicyclohexylamine, acmc-20ak4s, n,n-dicyclohexylethylamine
InChI Key XRKQMIFKHDXFNQ-UHFFFAOYSA-N
SMILES CCN(C1CCCCC1)C2CCCCC2
IUPAC Name N-cyclohexyl-N-ethylcyclohexanamine
Molecular Weight (g/mol) 209.38
PubChem CID 23563
Formula Weight 209.38
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material N-Ethyldicyclohexylamine, 96%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.