missing translation for 'onlineSavingsMsg'
Learn More

N-Ethyldeoxynojirimycin Hydrochloride, TRC

Product Code. 30372399
Change view
Click to view available options
Quantity:
5 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30372399 25 mg
30385422 5 mg
30375190 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30372399 Supplier Toronto Research Chemicals Supplier No. TRCE91500025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 210241-65-9
Molecular Formula C8 H17 N O4 . Cl H
Molecular Weight (g/mol) 227.69
IUPAC Name (2R,3R,4R,5S)-1-ethyl-2-(hydroxymethyl)piperidine-3,4,5-triol;hydrochloride
SMILES Cl.CCN1C[C@H](O)[C@@H](O)[C@H](O)[C@H]1CO

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C8H17NO4.ClH/c1-2-9-3-6(11)8(13)7(12)5(9)4-10;/h5-8,10-13H,2-4H2,1H3;1H/t5-,6+,7-,8-;/m1./s1
Formula Weight 227.0924
Chemical Name or Material N-Ethyldeoxynojirimycin, Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.