missing translation for 'onlineSavingsMsg'
Learn More

N-Ethylcarbamic Acid 3-[(1S)-1-(Dimethylamino)ethyl]phenyl Ester, TRC

Product Code. 30386510
Change view
Click to view available options
Quantity:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30386510 50 mg
30388012 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30386510 Supplier Toronto Research Chemicals Supplier No. TRCE90088550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1070660-34-2
Molecular Formula C13 H20 N2 O2
Molecular Weight (g/mol) 236.31
Synonym Carbamic acid, N-ethyl-, 3-[(1S)-1-(dimethylamino)ethyl]phenyl ester, 3-[(1S)-1-(Dimethylamino)ethyl]phenyl Ethyl Carbamate, N-Ethylcarbamic Acid 3-[(1S)-1-(Dimethylamino)ethyl]phenyl Ester
IUPAC Name [3-[(1S)-1-(dimethylamino)ethyl]phenyl] N-ethylcarbamate
SMILES CCNC(=O)Oc1cccc(c1)[C@H](C)N(C)C

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C13H20N2O2/c1-5-14-13(16)17-12-8-6-7-11(9-12)10(2)15(3)4/h6-10H,5H2,1-4H3,(H,14,16)/t10-/m0/s1
Formula Weight 236.1525
Chemical Name or Material N-Ethylcarbamic Acid 3-[(1S)-1-(Dimethylamino)ethyl]phenyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.