missing translation for 'onlineSavingsMsg'
Learn More

N-Ethylbenzylamine, 95%

Product Code. p-127597 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 10544721

Brand: Thermo Scientific 343710250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14321-27-8 | C9H13N | 135.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14321-27-8
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
MDL Number MFCD00009031
InChI Key HVAAHUDGWQAAOJ-UHFFFAOYSA-N
Synonym n-ethylbenzylamine, benzenemethanamine, n-ethyl, ethylbenzylamine, n-benzylethylamine, benzyl ethyl amine, benzylamine, n-ethyl, benzylethylamine, unii-mm8324ke8v, n-benzyl-n-ethylamine, benzyl-ethyl-amine
PubChem CID 84352
IUPAC Name N-benzylethanamine
SMILES CCNCC1=CC=CC=C1

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9000g/mL
Boiling Point 191.0°C to 194.0°C
Flash Point 66°C
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Linear Formula C6H5CH2NHC2H5
Refractive Index 1.5100 to 1.5130
Quantity 25 g
Beilstein 12,102
Specific Gravity 0.9
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in ethanol,ethyl ether and benzene
Formula Weight 135.21
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material N-Ethylbenzylamine, 95%
Show More Show Less
compliance-icons
Product Identifier
  • N-Ethylbenzylamine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.