missing translation for 'onlineSavingsMsg'
Learn More

N-Ethylaniline, 98%

Product Code. 10081080 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10081080 500 g Glass bottle 500g
10327130 100 g Glass bottle 100g
10306990 2.5 kg Glass bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10081080 Supplier Thermo Scientific Acros Supplier No. 118045000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 103-69-5 | C8H11N | 121.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-69-5
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.18
MDL Number MFCD00009025
InChI Key OJGMBLNIHDZDGS-UHFFFAOYSA-N
Synonym ethylaniline, n-ethyl aniline, benzenamine, n-ethyl, anilinoethane, n-ethylbenzenamine, ethylphenylamine, n-ethyl-n-phenylamine, aniline, n-ethyl, n-ethylaminobenzene, aethylanilin
PubChem CID 7670
ChEBI CHEBI:34870
IUPAC Name N-ethylaniline
SMILES CCNC1=CC=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -63°C
Color Brown to Yellow
Density 0.9630g/mL
Boiling Point 205°C
Flash Point 85°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5NHC2H5
Refractive Index 1.5530 to 1.555
Quantity 500 g
Beilstein 12, 159
Merck Index 15, 3819
Specific Gravity 0.963
Solubility Information Solubility in water: 50g/L water (20°C). Other solubilities: soluble in alcohol,ether and many other organic,solvents
Formula Weight 121.18
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material N-Ethylaniline, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • N-Ethylaniline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H373-May cause damage to organs.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.