missing translation for 'onlineSavingsMsg'
Learn More

N-Ethylaniline, 97%

Product Code. 11458750
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11458750 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11458750 Supplier Thermo Scientific Alfa Aesar Supplier No. A15060.AE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 103-69-5 | C8H11N | 121.18 g/mol

N-Ethylaniline (NEA) may be used as an internal standard in the GC analysis of nicotine extraction from nicotine gum. It may be used in the synthesis of the following, 2-(N-ethylphenylamino)-1,4-benzoquinone, 2-(arylaminomethyl)phenylboronic acid, poly(methyl methacrylate) (PMMA) films containing NEA and poly(N-ethylaniline) (PNEA).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Ethylaniline (NEA) may be used as an internal standard in the GC analysis of nicotine extraction from nicotine gum. It may be used in the synthesis of the following, 2-(N-ethylphenylamino)-1,4-benzoquinone, 2-(arylaminomethyl)phenylboronic acid, poly(methyl methacrylate) (PMMA) films containing NEA and poly(N-ethylaniline) (PNEA).

Solubility
Soluble in water (50g/L).

Notes
Air and Light Sensitive, store in dark and away from air. Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Specifications

CAS 103-69-5
Melting Point -64°C
Density 0.961
Boiling Point 204°C to 205°C
Flash Point 85°C
Odor Odorless
Molecular Formula C8H11N
Refractive Index 1.554
MDL Number MFCD00009025
Quantity 100 mL
UN Number UN2272
Beilstein 507468
Sensitivity Air and light sensitive
Merck Index 14,3764
Synonym ethylaniline, n-ethyl aniline, benzenamine, n-ethyl, anilinoethane, n-ethylbenzenamine
Solubility Information Soluble aq. soln. (50 g/L).
InChI Key OJGMBLNIHDZDGS-UHFFFAOYSA-N
SMILES CCNC1=CC=CC=C1
IUPAC Name N-ethylaniline
Molecular Weight (g/mol) 121.18
PubChem CID 7670
ChEBI CHEBI:34870
Formula Weight 121.18
Percent Purity 97%
Chemical Name or Material N-Ethylaniline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.