missing translation for 'onlineSavingsMsg'
Learn More

N'-Ethyl-N,N-dimethylethylenediamine, 98%

Product Code. 11362046
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11362046 25 g 25g
11352046 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11362046 Supplier Thermo Scientific Alfa Aesar Supplier No. L03180.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 123-83-1 | C6H18N2 | 118.22 g/mol

Reatant in the synthesis of tripod amphiphile. N'-Ethyl-N,N-dimethylethylenediamine is used as catalysts for olefin aziridination.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reatant in the synthesis of tripod amphiphile. N′-Ethyl-N,N-dimethylethylenediamine is used as catalysts for olefin aziridination.

Notes
Air Sensitive.Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air, carbondioxide.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-83-1
Molecular Formula C6H18N2
Molecular Weight (g/mol) 118.22
MDL Number MFCD00009034
InChI Key WLNSKTSWPYTNLY-UHFFFAOYSA-P
Synonym n,n-dimethyl-n'-ethylethylenediamine, 1,2-ethanediamine, n'-ethyl-n,n-dimethyl, 2-ethylaminoethyldimethylamine, 2-dimethylamino ethyl ethyl amine, ethylenediamine, n'-ethyl-n,n-dimethyl, n'-ethyl-n,n-dimethylethylenediamine, n1-ethyl-n2,n2-dimethylethane-1,2-diamine, 1,2-ethanediamine, n2-ethyl-n1,n1-dimethyl, n1-ethyl-n2,n2-dimethyl-1,2-ethanediamine, n'-ethyl-n,n-dimethylethane-1,2-diamine
PubChem CID 67162
IUPAC Name N-ethyl-N',N'-dimethylethane-1,2-diamine
SMILES CC[NH2+]CC[NH+](C)C

Specifications

Density 0.818
Boiling Point 134°C to 135°C
Flash Point 23°C (73°F)
Odor Amine-like
Refractive Index 1.4225
Quantity 25 g
UN Number UN2734
Beilstein 773675
Formula Weight 116.21
Percent Purity 98%
Chemical Name or Material N'-Ethyl-N,N-dimethylethylenediamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.