missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Ethyl-N-methylbenzylamine, 99%

Product Code. 11388205
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11388205 10 g 10g
11398205 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11388205 Supplier Alfa Aesar™ Supplier No. L08520.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 4788-37-8
Molecular Formula C10H15N
Molecular Weight (g/mol) 149.237
MDL Number MFCD00051914
InChI Key ABRGFZIXRKAUJS-UHFFFAOYSA-N
Synonym n-ethyl-n-methylbenzylamine, n-benzyl-n-ethylmethylamine, benzyl ethyl methylamine, ethylmethylbenzylamine, benzylethylmethylamine, acmc-209kat, benzyl-ethyl-methyl-amine, s-methylethylbenzylamine, n-methyl-n-ethyl-n-benzylamine
PubChem CID 248797
IUPAC Name N-benzyl-N-methylethanamine
SMILES CCN(C)CC1=CC=CC=C1

Specifications

Density 1.03
Boiling Point 196°C
Refractive Index 1.4995
Quantity 10 g
UN Number UN2735
Beilstein 2082378
Formula Weight 149.24
Percent Purity 99%
Chemical Name or Material N-Ethyl-N-methylbenzylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.