missing translation for 'onlineSavingsMsg'
Learn More

N-Ethyl-N-(4-methylbenzyl)-3-nitrobenzenesulfonamide, 97%, Thermo Scientific™

Catalog Number 15434626
Click to view available options
Quantity:
100 mg
500 mg
This item is not returnable. View return policy
1421263-31-1
C16H18N2O4S
334.39
MFCD22683023
YESMQOFCWVUVNT-UHFFFAOYSA-N
n-ethyl-n-4-methylphenyl methyl-3-nitrobenzenesulfonamide, n-ethyl-n-4-methylbenzyl-3-nitrobenzenesulfonamide
97290227
N-ethyl-N-[(4-methylphenyl)methyl]-3-nitrobenzenesulfonamide
CCN(CC1=CC=C(C)C=C1)S(=O)(=O)C1=CC=CC(=C1)[N+]([O-])=O
This item is not returnable. View return policy
1421263-31-1
C16H18N2O4S
334.39
MFCD22683023
YESMQOFCWVUVNT-UHFFFAOYSA-N
n-ethyl-n-4-methylphenyl methyl-3-nitrobenzenesulfonamide, n-ethyl-n-4-methylbenzyl-3-nitrobenzenesulfonamide
97290227
N-ethyl-N-[(4-methylphenyl)methyl]-3-nitrobenzenesulfonamide
CCN(CC1=CC=C(C)C=C1)S(=O)(=O)C1=CC=CC(=C1)[N+]([O-])=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 100 mg
Formula Weight 334.38
Percent Purity 97%
Chemical Name or Material N-Ethyl-N-(4-methylbenzyl)-3-nitrobenzenesulfonamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'